siRNA Details
| Gene Symbol: Gm12002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040873.1 | - | 5 | 501 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK796166.1 | - | 5 | 508 | Chr. 11 - 12314532 - 12336060 [+] on Build GRCm38 | |

Gm12002
Genomic Map