siRNA Details
| Gene Symbol: Ttc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005733.1 | - | 2 | 311 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705898.1 | - | 2 | 306 | Chr. 17 - 87329789 - 87332049 [-] on Build GRCm38 | |

Ttc7
Genomic Map