siRNA Details
| Gene Symbol: Gm11217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076794.1 | - | 4 | 341 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615287.1 | - | 4 | 338 | Chr. 4 - 64255101 - 64276601 [-] on Build GRCm38 | |

Gm11217
Genomic Map