siRNA Details
| Gene Symbol: Gm32754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076796.1 | - | 29 | |||
| Gene Symbol: Tmem232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF338468.1 | - | 1 | 33 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615300.1 | - | 20 | Chr. 3 - 140905191 - 140917459 [-] on Build GRCm38 | ||

Gm32754
Genomic Map