siRNA Details
| Gene Symbol: Gm32754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076796.1 | - | 788 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB013538.2 | - | 4 | 608 | Chr. 3 - 140904998 - 140917352 [-] on Build GRCm38 | |

Gm32754
Genomic Map