siRNA Details
| Gene Symbol: A930033M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA547551.1 | - | 1 | 114 | ||
| AI504129.1 | - | 1 | 121 | |||
| AK020925.1 | - | 1 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930033M14Rik| Gene Symbol: A930033M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA547551.1 | - | 1 | 114 | ||
| AI504129.1 | - | 1 | 121 | |||
| AK020925.1 | - | 1 | 140 | |||