siRNA Details
| Gene Symbol: A930033M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA547551.1 | - | 1 | 137 | ||
| AI504129.1 | - | 1 | 144 | |||
| AK020925.1 | - | 1 | 163 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB284009.1 | - | 1 | 174 | Chr. 10 - 40260945 - 40261333 [+] on Build GRCm38 | |

A930033M14Rik
Genomic Map