siRNA Details
| Gene Symbol: Gm5563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087181.1 | - | 2 | 621 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB533934.2 | - | 1 | 538 | Chr. 5 - 119830946 - 119831596 [-] on Build GRCm38 | |

Gm5563
Genomic Map