siRNA Details
| Gene Symbol: Gm33337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089123.1 | - | 2 | 1022 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB559987.2 | - | 2 | 576 | Chr. 10 - 118718146 - 118718981 [-] on Build GRCm38 | |

Gm33337
Genomic Map