siRNA Details
| Gene Symbol: 8430426J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077230.1 | 1 | 78 | |||
| GenBank mRNA | BC038155.1 | - | 1 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430426J06Rik| Gene Symbol: 8430426J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077230.1 | 1 | 78 | |||
| GenBank mRNA | BC038155.1 | - | 1 | 78 | ||