siRNA Details
| Gene Symbol: Klhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033046.1 | - | 2548 | |||
| AK082138.1 | - | 427 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX238931.1 | - | 1 | 197 | Chr. 13 - 58000808 - 58001643 [-] on Build GRCm38 | |

Klhl3
Genomic Map