siRNA Details
| Gene Symbol: Klhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033046.1 | - | 2154 | |||
| AK082138.1 | - | 33 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl3| Gene Symbol: Klhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033046.1 | - | 2154 | |||
| AK082138.1 | - | 33 | ||||