siRNA Details
| Gene Symbol: 1700019L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006131.1 | - | 4 | 560 | ||
| AV209170.1 | - | 3 | 270 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706236.1 | - | 4 | 557 | Chr. 8 - 50392710 - 50517639 [-] on Build GRCm38 | |

1700019L22Rik
Genomic Map