siRNA Details
Gene Symbol: 4930428E07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045943.1 | 3 | 315 | |||
GenBank mRNA | AK015219.1 | - | 3 | 315 | ||
AK030072.1 | - | 3 | 316 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930428E07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045943.1 | 3 | 315 | |||
GenBank mRNA | AK015219.1 | - | 3 | 315 | ||
AK030072.1 | - | 3 | 316 |