siRNA Details
| Gene Symbol: 4930428E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045943.1 | 2, 3 | 254 | |||
| GenBank mRNA | AK015219.1 | - | 2, 3 | 254 | ||
| AK030072.1 | - | 2, 3 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930428E07Rik| Gene Symbol: 4930428E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045943.1 | 2, 3 | 254 | |||
| GenBank mRNA | AK015219.1 | - | 2, 3 | 254 | ||
| AK030072.1 | - | 2, 3 | 255 | |||