siRNA Details
| Gene Symbol: 4930428E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045943.1 | 9 | 892 | |||
| GenBank mRNA | AK015219.1 | - | 9 | 892 | ||
| AK030072.1 | - | 5 | 3787 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV262118.1 | - | 4 | 226 | Chr. 12 - 38552808 - 38560978 [-] on Build GRCm38 | |

4930428E07Rik
Genomic Map