siRNA Details
| Gene Symbol: 4931406E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016433.1 | - | 1 | 31 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716088.1 | - | 1 | 29 | Chr. 11 - 88396515 - 88397527 [+] on Build GRCm38 | |

4931406E20Rik
Genomic Map