siRNA Details
| Gene Symbol: Gm2511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC100510.1 | - | 4 | 385 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN836674.1 | - | 388 | Chr. - - [-] on Build | ||

Gm2511
Genomic Map