siRNA Details
| Gene Symbol: Gm15651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085357.1 | - | 2 | 1907 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB498603.2 | - | 1 | 33 | Chr. 16 - 22550514 - 22551146 [+] on Build GRCm38 | |

Gm15651
Genomic Map