siRNA Details
| Gene Symbol: Gm34517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132743.1 | - | 1 | 311 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ046802.1 | - | 1 | 311 | Chr. 19 - 43266936 - 43277499 [+] on Build GRCm38 | |

Gm34517
Genomic Map