siRNA Details
| Gene Symbol: Gm39091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080247.1 | - | 3 | 392 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB637665.1 | - | 3 | 390 | Chr. 7 - 128182379 - 128184096 [-] on Build GRCm38 | |

Gm39091
Genomic Map