siRNA Details
| Gene Symbol: Car7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038270.1 | - | 1 | 94 | ||
| BY741075.1 | - | 1 | 119 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK148814.1 | - | 132 | Chr. - - [-] on Build | ||

Car7
Genomic Map