siRNA Details
| Gene Symbol: Sstr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051189.1 | - | 1539 | |||
| AK158660.1 | - | 206 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ144546.1 | - | 1 | 206 | Chr. 2 - 148399847 - 148400399 [+] on Build GRCm38 | |

Sstr4
Genomic Map