siRNA Details
| Gene Symbol: Gm7596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145172.1 | - | 341 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY735114.1 | - | 2 | 338 | Chr. 5 - 32390173 - 32393303 [-] on Build GRCm38 | |

Gm7596
Genomic Map