siRNA Details
| Gene Symbol: Gm34292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142341.1 | - | 1 | 23 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730467.1 | - | 1 | 23 | Chr. 2 - 144555696 - 144556155 [-] on Build GRCm38 | |

Gm34292
Genomic Map