siRNA Details
| Gene Symbol: Pon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018124.1 | - | 1285 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB672064.1 | - | 1 | 65 | Chr. 6 - 5263212 - 5263631 [-] on Build GRCm38 | |

Pon2
Genomic Map