siRNA Details
Gene Symbol: 8430422H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045373.1 | 4 | 1150 | |||
GenBank mRNA | AK018433.1 | - | 4 | 1153 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 8430422H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045373.1 | 4 | 1150 | |||
GenBank mRNA | AK018433.1 | - | 4 | 1153 |