siRNA Details
| Gene Symbol: 2500004C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040318.1 | 2 | 1724 | |||
| GenBank mRNA | AK010870.1 | - | 2 | 1636 | ||
| AV103564.1 | - | 1 | 118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2500004C02Rik| Gene Symbol: 2500004C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040318.1 | 2 | 1724 | |||
| GenBank mRNA | AK010870.1 | - | 2 | 1636 | ||
| AV103564.1 | - | 1 | 118 | |||