siRNA Details
| Gene Symbol: 1700018A23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006078.1 | - | 2 | 193 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706191.1 | - | 2 | 193 | Chr. 6 - 126822377 - 126825177 [+] on Build GRCm38 | |

1700018A23Rik
Genomic Map