siRNA Details
| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029820.1 | - | 3259 | |||
| AK037996.1 | - | 2238 | ||||
| AK078985.1 | - | 380 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ldhd| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029820.1 | - | 3259 | |||
| AK037996.1 | - | 2238 | ||||
| AK078985.1 | - | 380 | ||||