siRNA Details
Gene Symbol: 9430019H13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020425.1 | - | 1 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9430019H13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020425.1 | - | 1 | 88 |