siRNA Details
| Gene Symbol: R3hcc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006610.1 | - | 2 | 192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

R3hcc1l| Gene Symbol: R3hcc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006610.1 | - | 2 | 192 | ||