siRNA Details
| Gene Symbol: 1700037C06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006618.1 | - | 1 | 851 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF452515.1 | - | 1 | 289 | Chr. 4 - 137116705 - 137117022 [-] on Build GRCm38 | |
 Back To Top
 Back To Top
 1700037C06Rik
1700037C06Rik Genomic Map
Genomic Map Back To Top
 Back To Top