siRNA Details
| Gene Symbol: 1700037C06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006618.1 | - | 1 | 217 | ||
| Gene Symbol: Gm13001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CA465487.1 | - | 2 | 479 | ||

1700037C06Rik
Genomic Map