siRNA Details
| Gene Symbol: 1700037C06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006618.1 | - | 1 | 782 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF452515.1 | - | 1 | 220 | Chr. 4 - 137116705 - 137117022 [-] on Build GRCm38 | |

1700037C06Rik
Genomic Map