siRNA Details
Gene Symbol: Redrum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040338.1 | 1 | 128 | |||
GenBank mRNA | AK036466.1 | - | 1 | 129 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Redrum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040338.1 | 1 | 128 | |||
GenBank mRNA | AK036466.1 | - | 1 | 129 |