siRNA Details
| Gene Symbol: Gm35073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145828.1 | - | 700 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY757141.1 | - | 1 | 576 | Chr. 4 - 98699854 - 98700531 [-] on Build GRCm38 | |

Gm35073
Genomic Map