siRNA Details
| Gene Symbol: 4933406C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_044986.1 | 6 | 1017 | |||
| GenBank mRNA | AK041289.1 | - | 6 | 1018 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933406C10Rik| Gene Symbol: 4933406C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_044986.1 | 6 | 1017 | |||
| GenBank mRNA | AK041289.1 | - | 6 | 1018 | ||