siRNA Details
| Gene Symbol: 2810417H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134432.1 | - | 2 | 551 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY748272.1 | - | 2 | 549 | Chr. 9 - 65890362 - 65891275 [+] on Build GRCm38 | |

2810417H13Rik
Genomic Map