siRNA Details
| Gene Symbol: 1700074A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006945.1 | - | 1 | 308 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706969.1 | - | 1 | 306 | Chr. 2 - 181837417 - 181837798 [-] on Build GRCm38 | |

1700074A11Rik
Genomic Map