siRNA Details
| Gene Symbol: 4930466I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015514.1 | - | 2 | 1737 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930466I24Rik| Gene Symbol: 4930466I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015514.1 | - | 2 | 1737 | ||