siRNA Details
| Gene Symbol: 4930466I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015514.1 | - | 2 | 428 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715061.1 | - | 2 | 425 | Chr. 6 - 92013766 - 92016115 [-] on Build GRCm38 | |

4930466I24Rik
Genomic Map