siRNA Details
| Gene Symbol: 4930466K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015515.1 | - | 1 | 496 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU610016.1 | - | 1 | 580 | Chr. 10 - 53614284 - 53614999 [+] on Build GRCm38 | |

4930466K18Rik
Genomic Map