siRNA Details
| Gene Symbol: Kdm7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK166173.1 | - | 1 | 1644 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG803042.1 | - | 1 | 163 | Chr. 6 - 39153346 - 39153944 [-] on Build GRCm38 | |

Kdm7a
Genomic Map