siRNA Details
Gene Symbol: C030029H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102277.1 | 3, 4 | 1485 | |||
GenBank mRNA | AK034027.1 | - | 3, 4 | 1487 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C030029H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102277.1 | 3, 4 | 1485 | |||
GenBank mRNA | AK034027.1 | - | 3, 4 | 1487 |