siRNA Details
| Gene Symbol: C030029H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102277.1 | 4 | 1898 | |||
| GenBank mRNA | AK034027.1 | - | 4 | 1900 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030029H02Rik| Gene Symbol: C030029H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102277.1 | 4 | 1898 | |||
| GenBank mRNA | AK034027.1 | - | 4 | 1900 | ||