siRNA Details
| Gene Symbol: Gm5547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045845.1 | 2 | 67 | |||
| GenBank mRNA | AK138691.1 | - | 2 | 67 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY724460.1 | - | 2 | 63 | Chr. 3 - 105815571 - 105817349 [+] on Build GRCm38 | |

Gm5547
Genomic Map