siRNA Details
| Gene Symbol: 1700074G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018899.1 | - | 1 | 127 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706975.1 | - | 1 | 127 | Chr. 11 - 116589621 - 116589781 [-] on Build GRCm38 | |

1700074G04Rik
Genomic Map