siRNA Details
| Gene Symbol: Gm11351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045062.1 | 1, 2 | 42 | |||
| GenBank mRNA | AK132076.1 | - | 1, 2 | 42 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712905.1 | - | 1, 2 | 42 | Chr. 13 - 25921070 - 25966316 [-] on Build GRCm38 | |

Gm11351
Genomic Map