siRNA Details
| Gene Symbol: Gm28722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132070.1 | - | 1 | 60 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712897.1 | - | 1 | 60 | Chr. 1 - 90453733 - 90454501 [-] on Build GRCm38 | |

Gm28722
Genomic Map